Mouse IgG1 Isotype Control [clone: MOPC31C] (MOPC31C)

Supplier: Adipogen
ANC-278-010
102512-774EA 222.92 USD
102512-774
Mouse IgG1 Isotype Control [clone: MOPC31C] (MOPC31C)
Antibodies
Immunoglobulin expressed by the mouse plasmacytoma MOPC 31C is an IgG1kappa isotype of unknown specificity. It is used as a negative control when working with human cells.

Type: Control
Antigen:
Clonality: Monoclonal
Clone: MOPC31C
Conjugation: Unconjugated
Epitope:
Host:
Isotype: Mouse IgG1kappa
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR