Anti-CD268 Monoclonal Antibody [clone: ANC268.2/6E6]

Supplier: Adipogen
ANC-275-020
102512-768EA 350.31 USD
102512-768
Anti-CD268 Monoclonal Antibody [clone: ANC268.2/6E6]
Antibodies
Three receptors for BAFF were recently identified and designated TACI, BCMA and BAFF-R. BAFF-R signaling is essential for B cell development, survival and humoral immune responses.

Type: Primary
Antigen: CD268
Clonality: Monoclonal
Clone: ANC268.2/6E6
Conjugation: Unconjugated
Epitope:
Host:
Isotype: Mouse IgG1kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR