Anti-CD270 Monoclonal Antibody (R-PE) [clone: ANC3B7]

Supplier: Adipogen
ANC-270-050
102512-746EA 520.62 USD
102512-746
Anti-CD270 Monoclonal Antibody (R-PE) [clone: ANC3B7]
Antibodies
Human CD270 (Herpes Virus Entry Mediator; TNFRSF14; ATAR; LIGHTR; TR2) is a 36kDa type I membrane protein that has been found on a broad range of lymphoid tissues. Its ligands include the TNF ligand family members LTalpha and LIGHT and the CD28/Ig superfamily member BTLA.

Type: Primary
Antigen: CD270
Clonality: Monoclonal
Clone: ANC3B7
Conjugation: R-PE
Epitope:
Host:
Isotype: Mouse IgG2akappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR