Anti-LTBETAR Monoclonal Antibody (Biotin) [clone: ANCLTR2/9E2]

Supplier: Adipogen
ANC-267-030
102512-724EA 483.23 USD
102512-724
Anti-LTBETAR Monoclonal Antibody (Biotin) [clone: ANCLTR2/9E2]
Antibodies
The LTbetaR activates two different NF-kappaB pathways that lead to distinct patterns of gene induction, including selected chemokines and the cytokine BAFF, which is essential for the survival of mature B lymphocytes. LTbetaR activates the classical NF-kappaB (relA/p50) pathway, like the type 1 TNF receptor (TNFR1), that regulates proinflammatory genes, like the chemokine MIP1beta. However, LTbetaR, unlike TNFR1, also activates the processing of p100 to form RelB/p52 complexes, which activate genes involved in lymphoid organ formation and lymphocyte survival.

Type: Primary
Antigen: LTBETAR
Clonality: Monoclonal
Clone: ANCLTR2/9E2
Conjugation: Biotin
Epitope:
Host:
Isotype: Mouse IgG1kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR