Anti-CD278 Monoclonal Antibody (R-PE) [clone: ANC6C6]

Supplier: Adipogen
ANC-265-050
102512-708EA 520.62 USD
102512-708
Anti-CD278 Monoclonal Antibody (R-PE) [clone: ANC6C6]
Antibodies
The inducible costimulator CD278 (ICOS) is similar to human CD28 (24% homology) and plays an analogous role in the T cell activation process. Secondary signaling through CD28 or CD278 results in discrete cytokine secretion profiles by the activated T cells. CD275 (ICOSL; GL50; B7-H2) has been shown to be the ligand for CD278 (ICOS).

Type: Primary
Antigen: CD278
Clonality: Monoclonal
Clone: ANC6C6
Conjugation: R-PE
Epitope:
Host:
Isotype: Mouse IgG1kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR