Anti-IFNG Monoclonal Antibody (FITC) [clone: ANC2E11]

Supplier: Adipogen
ANC-247-040
102512-640EA 350.31 USD
102512-640
Anti-IFNG Monoclonal Antibody (FITC) [clone: ANC2E11]
Antibodies
The cytokine human interferon-gamma (IFN-gamma) is a glycosylated protein that exists as a homodimer of 34kDa in its biologically active form. IFN-gamma is expressed upon activation by TH1 cells CD8+ cells and NK cells. Secreted IFN-gamma is a potent inhibitor of in vitro hematopoiesis and appears to serve as an important regulator of lymphocyte and macrophage function by exerting antiproliferative effects.

Type: Primary
Antigen: IFNG
Clonality: Monoclonal
Clone: ANC2E11
Conjugation: FITC
Epitope:
Host:
Isotype: Mouse IgG1kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR