Anti-IL-2 Monoclonal Antibody (Biotin) [clone: ANC7F7]

Supplier: Adipogen
ANC-245-030
102512-630EA 350.31 USD
102512-630
Anti-IL-2 Monoclonal Antibody (Biotin) [clone: ANC7F7]
Antibodies
The cytokine human interleukin-2 (IL-2) is a 15kDa protein expressed upon activation by peripheral TH1 cells, medullary thymocytes and a subset of large granular lymphocytes. Secreted IL-2 stimulates proliferation and lymphokine production by T cells, B cells and NK cells.

Type: Primary
Antigen: IL-2
Clonality: Monoclonal
Clone: ANC7F7
Conjugation: Biotin
Epitope:
Host:
Isotype: Mouse IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR