Anti-CD62E Monoclonal Antibody (R-PE) [clone: HAE-1f]

Supplier: Adipogen
ANC-240-050
102512-626EA 447.23 USD
102512-626
Anti-CD62E Monoclonal Antibody (R-PE) [clone: HAE-1f]
Antibodies
Human CD62E (E-selectin) is an adhesion molecule expressed on activated endothelium and facilitates recruitment of leukocytes to sights of acute inflammation and immunological events. CD62E has affinity to Lewisx.

Type: Primary
Antigen: CD62E
Clonality: Monoclonal
Clone: HAE-1f
Conjugation: R-PE
Epitope:
Host:
Isotype: Mouse IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR