Anti-CD79A Monoclonal Antibody [clone: ZL7-4]

Supplier: Adipogen
ANC-235-020
102512-608EA 297.69 USD
102512-608
Anti-CD79A Monoclonal Antibody [clone: ZL7-4]
Antibodies
Human CD79a is a 22kDa glycoprotein which associates with CD79b to form a functional heterodimer that interacts with membrane immunoglobulin (mIg).The mIgM component binds antigen and the heterodimer transduces signals.

Type: Primary
Antigen: CD79A
Clonality: Monoclonal
Clone: ZL7-4
Conjugation: Unconjugated
Epitope:
Host:
Isotype: Mouse IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR