Anti-IgG/IgM Goat Polyclonal Antibody (Biotin)

Supplier: Adipogen
ANC-234-011
102512-606EA 350.31 USD
102512-606
Anti-IgG/IgM Goat Polyclonal Antibody (Biotin)
Antibodies
Goat anti-mouse/FITC is reactive with mouse Ig subclasses: G1, G2a, G2b, G3. It also reacts well with mouse IgM antibodies. Reagent does not bind significantly to any human cells or cell lines tested.

Type: Secondary
Antigen: IgG/IgM
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Goat
Isotype:
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR