Anti-CD75 Monoclonal Antibody [clone: LN1]

Supplier: Adipogen
ANC-228-020
102512-592EA 297.69 USD
102512-592
Anti-CD75 Monoclonal Antibody [clone: LN1]
Antibodies
Human CD75 is an alpha 2, 6-sialylated carbohydrate molecule found mainly on mature B cells and germinal center B cells and also in T cell subsets and erythrocitic cells. CD75 appears to be a ligand for CD22.

Type: Primary
Antigen: CD75
Clonality: Monoclonal
Clone: LN1
Conjugation: Unconjugated
Epitope:
Host:
Isotype: Mouse IgM
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR