Anti-CD70 Monoclonal Antibody (R-PE) [clone: BU69]

Supplier: Adipogen
ANC-222-050
102512-562EA 447.23 USD
102512-562
Anti-CD70 Monoclonal Antibody (R-PE) [clone: BU69]
Antibodies
Human CD70 is not present on resting lymphocytes. CD70 is expressed after activation on about 20% of T cells and on about 70% of B cells. CD70 is a ligand for CD27 and it is also a member of the TNF gene family.

Type: Primary
Antigen: CD70
Clonality: Monoclonal
Clone: BU69
Conjugation: R-PE
Epitope:
Host:
Isotype: Mouse IgG1
Reactivity: Human, Primate
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR