Anti-CD69 Monoclonal Antibody [clone: HP-4B3]

Supplier: Adipogen
ANC-221-020
102512-544EA 297.69 USD
102512-544
Anti-CD69 Monoclonal Antibody [clone: HP-4B3]
Antibodies
Human CD69 is expressed by platelets and mature thymocytes and appears rapidly after activation on T cells, B cells and NK cells. This pattern of cellular CD69 expression has suggested a lymphocyte activation role. Functional studies with antibodies to CD69 also support an activation role for CD69.

Type: Primary
Antigen: CD69
Clonality: Monoclonal
Clone: HP-4B3
Conjugation: Unconjugated
Epitope:
Host:
Isotype: Mouse IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR