Anti-CD63 Monoclonal Antibody (Biotin) [clone: AHN16.1]

Supplier: Adipogen
ANC-215-030
102512-516EA 447.23 USD
102512-516
Anti-CD63 Monoclonal Antibody (Biotin) [clone: AHN16.1]
Antibodies
Human CD63 is expressed on activated platelets and also on monocytes and macrophages. CD63 contains four hydrophobic transmembrane regions and is a member of the tetraspan family. CD63 appears to be involved with cell adhesion and associates with VLA-3 and VLA-6 integrins. In a colon cancer model, it has been correlated to metastatic potential.

Type: Primary
Antigen: CD63
Clonality: Monoclonal
Clone: AHN16.1
Conjugation: Biotin
Epitope:
Host:
Isotype: Mouse IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR