Anti-CD59 Monoclonal Antibody [clone: BRA-10G]

Supplier: Adipogen
ANC-211-020
102512-494EA 297.69 USD
102512-494
Anti-CD59 Monoclonal Antibody [clone: BRA-10G]
Antibodies
Human CD59 is a GPI anchored molecule that protects cells from complement-mediated lysis. Studies are progressing to determine the regulatory mechanism for CD59 expression. A CD59-like protein has been discovered on the causative agent of Amebic encephalitis.

Type: Primary
Antigen: CD59
Clonality: Monoclonal
Clone: BRA-10G
Conjugation: Unconjugated
Epitope:
Host:
Isotype: Mouse IgG1kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR