Anti-CD58 Monoclonal Antibody (Biotin) [clone: TS2]

Supplier: Adipogen
ANC-210-030
102512-488EA 350.31 USD
102512-488
Anti-CD58 Monoclonal Antibody (Biotin) [clone: TS2]
Antibodies
Human CD58 is a cell surface glycoprotein with wide distribution on hemopoietic cells and functions as an adhesion molecule mediating a costimulatory signal through CD2. CD58 is attached to the cell surface by either of two forms - transmembrane or GPI.

Type: Primary
Antigen: CD58
Clonality: Monoclonal
Clone: TS2
Conjugation: Biotin
Epitope:
Host:
Isotype: Mouse IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR