Anti-CD57 Monoclonal Antibody [clone: NK-1]

Supplier: Adipogen
ANC-209-020
102512-476EA 297.69 USD
102512-476
Anti-CD57 Monoclonal Antibody [clone: NK-1]
Antibodies
Human CD57 originally called HNK-1, is a glycoprotein found on 15-20 % of PBL’s (peripheral blood lymphocytes) including 60 % of NK cells and a subset of T cells. The immune regulation role of CD57 positive PBL’s expressing high levels of CD8 is being investigated.

Type: Primary
Antigen: CD57
Clonality: Monoclonal
Clone: NK-1
Conjugation: Unconjugated
Epitope:
Host:
Isotype: Mouse IgM
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR