Anti-CD56 Monoclonal Antibody (Biotin) [clone: ERIC-1]

Supplier: Adipogen
ANC-208-030
102512-474EA 447.23 USD
102512-474
Anti-CD56 Monoclonal Antibody (Biotin) [clone: ERIC-1]
Antibodies
Human CD56 is an adhesion molecule from the Ig superfamily which is restricted to NK cells in the immune system. It is believed that NK cells form a first line of defense against tumor cells and cells infected with bacteria and viruses.

Type: Primary
Antigen: CD56
Clonality: Monoclonal
Clone: ERIC-1
Conjugation: Biotin
Epitope:
Host:
Isotype: Mouse IgG1kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR