Anti-CD55 Monoclonal Antibody (Biotin) [clone: 67]

Supplier: Adipogen
ANC-207-030
102512-464EA 350.31 USD
102512-464
Anti-CD55 Monoclonal Antibody (Biotin) [clone: 67]
Antibodies
Human CD55 is a GPI anchored protein that can protect cells from complement mediated lysis.

Type: Primary
Antigen: CD55
Clonality: Monoclonal
Clone: 67
Conjugation: Biotin
Epitope:
Host:
Isotype: Mouse IgG1kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR