Anti-CD54 Monoclonal Antibody [clone: 15.2]

Supplier: Adipogen
ANC-205-020
102512-440EA 297.69 USD
102512-440
Anti-CD54 Monoclonal Antibody [clone: 15.2]
Antibodies
Human CD54 (ICAM-1) mediates cell adhesion by binding to the integrins CD11a/CD18 (LFA-1) and to CD11b/CD18 (Mac-1). CD54 expression on resting peripheral blood leukocytes is weak but is upregulated on activated T and B lymphocytes and on monocytes.

Type: Primary
Antigen: CD54
Clonality: Monoclonal
Clone: 15.2
Conjugation: Unconjugated
Epitope:
Host:
Isotype: Mouse IgG1kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR