Anti-CD53 Monoclonal Antibody (FITC) [clone: 63.5A3]

Supplier: Adipogen
ANC-204-040
102512-436EA 350.31 USD
102512-436
Anti-CD53 Monoclonal Antibody (FITC) [clone: 63.5A3]
Antibodies
Human CD53 is a tetraspan cell surface glycoprotein found on all hemopoetic cells except platelets and red blood cells.

Type: Primary
Antigen: CD53
Clonality: Monoclonal
Clone: 63.5A3
Conjugation: FITC
Epitope:
Host:
Isotype: Mouse IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR