Anti-CD51 Monoclonal Antibody (R-PE) [clone: P2W7]

Supplier: Adipogen
ANC-202-050
102512-428EA 447.23 USD
102512-428
Anti-CD51 Monoclonal Antibody (R-PE) [clone: P2W7]
Antibodies
Human CD51 is an integrin alpha v chain which can associate with five different integrin beta chains to form various adhesion molecules. The alpha v beta 3 integrin complex binds vitronectin at the RGD sequence and also can bind to fibrinogen, von Willebrand factor, thrombospondin, fibronectin, osteopontin and collagen.

Type: Primary
Antigen: CD51
Clonality: Monoclonal
Clone: P2W7
Conjugation: R-PE
Epitope:
Host:
Isotype: Mouse IgG1kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR