Anti-CD49d Monoclonal Antibody (Biotin) [clone: BU49]

Supplier: Adipogen
ANC-200-030
102512-404EA 350.31 USD
102512-404
Anti-CD49d Monoclonal Antibody (Biotin) [clone: BU49]
Antibodies
Human CD49d molecules are integrin alpha4 chains that are expressed as heterodimers with CD29 (beta 1 integrin) or (beta 7 integrin) and function as adhesion receptors. CD49d/CD29 is mainly expressed on thymocytes and B cells with increased expression on activated T cells. The ligands for CD49d/CD29 include VCAM-1 and fibronectin.

Type: Primary
Antigen: CD49d
Clonality: Monoclonal
Clone: BU49
Conjugation: Biotin
Epitope:
Host:
Isotype: Mouse IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR