Anti-CD45 Monoclonal Antibody (FITC) [clone: C11]

Supplier: Adipogen
ANC-196-040
102512-376EA 350.31 USD
102512-376
Anti-CD45 Monoclonal Antibody (FITC) [clone: C11]
Antibodies
Human CD45 is expressed on all cells of hematopoietic origin, except erythrocytes. CD45 is a transmembrane tyrosine phosphatase which can exist in at least nine different isoforms resulting from tissue-specific alternative RNA splicing of exons 4-7 of a single gene coding for the various N-terminal peptide segments. CD45 plays an essential role in lymphocyte activation.

Type: Primary
Antigen: CD45
Clonality: Monoclonal
Clone: C11
Conjugation: FITC
Epitope:
Host:
Isotype: Mouse IgG2akappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR