Anti-CD45RO Monoclonal Antibody (R-PE) [clone: UCHL1]

Supplier: Adipogen
ANC-194-050
102512-360EA 447.23 USD
102512-360
Anti-CD45RO Monoclonal Antibody (R-PE) [clone: UCHL1]
Antibodies
Human CD45 is expressed on all cells of hematopoietic origin, except erythrocytes. CD45 is a transmembrane tyrosine phosphatase which can exist in at least nine different isoforms resulting from tissue-specific alternative RNA splicing of exons 4-7 of a single gene coding for the various N-terminal peptide segments. The CD45RO isoform contains none of the alternatively spliced exons and is present on most thymocytes, a subpopulation of resting (memory) T cells within both CD4 and CD8 subsets as well as on monocytes and granulocytes. CD45RO plays an essential role in lymphocyte activation.

Type: Primary
Antigen: CD45RO
Clonality: Monoclonal
Clone: UCHL1
Conjugation: R-PE
Epitope:
Host:
Isotype: Mouse IgG2a
Reactivity: Human, Primate
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR