Anti-CD43 Monoclonal Antibody [clone: DFT1]

Supplier: Adipogen
ANC-192-820
102512-332EA 297.69 USD
102512-332
Anti-CD43 Monoclonal Antibody [clone: DFT1]
Antibodies
Human CD43 is a heavily O-glycosylated and sialylated transmembrane protein that is mainly found on T cells, but not on B cells.

  • preservative free

Type: Primary
Antigen: CD43
Clonality: Monoclonal
Clone: DFT1
Conjugation: Preservative Free
Epitope:
Host:
Isotype: Mouse IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR