Anti-CD41a Monoclonal Antibody (Biotin) [clone: 96.2C1]

Supplier: Adipogen
ANC-190-030
102512-324EA 350.31 USD
102512-324
Anti-CD41a Monoclonal Antibody (Biotin) [clone: 96.2C1]
Antibodies
Human CD41 is a member of a major platelet transmembrane glycoprotein complex (CD41/CD61) which mediates platelet aggregation and adhesion by serving as the receptor for fibrinogen and von Willebrand factor, fibronectin and vitronectin.

Type: Primary
Antigen: CD41a
Clonality: Monoclonal
Clone: 96.2C1
Conjugation: Biotin
Epitope:
Host:
Isotype: Mouse IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR