Anti-CD40 Monoclonal Antibody [clone: BE-1]

Supplier: Adipogen
ANC-189-820
102512-312EA 297.69 USD
102512-312
Anti-CD40 Monoclonal Antibody [clone: BE-1]
Antibodies
Human CD40 is a member of the TNF receptor family and is present on all B cells except plasma cells. CD40 is also found on some epithelial cells, carcinomas and lymphoid dendritic cells. CD40 plays an important role in B cell activation and the interaction with its ligand CD154 (CD40 ligand) is essential for isotype switching.

  • preservative free

Type: Primary
Antigen: CD40
Clonality: Monoclonal
Clone: BE-1
Conjugation: Preservative Free
Epitope:
Host:
Isotype: Mouse IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR