Anti-CD39 Monoclonal Antibody (Biotin) [clone: BU61]

Supplier: Adipogen
ANC-188-030
102512-304EA 350.31 USD
102512-304
Anti-CD39 Monoclonal Antibody (Biotin) [clone: BU61]
Antibodies
Human CD39 is found on most mature B cells, activated NK cells and activated T cells, as well as neutrophills and monocytes. CD39 has homology to the N-myc family of proteins and has been cloned. CD39 is an ecto apyrase. A lower expression level of CD39 on T and NK cells have been observed in hypertensive individuals.

Type: Primary
Antigen: CD39
Clonality: Monoclonal
Clone: BU61
Conjugation: Biotin
Epitope:
Host:
Isotype: Mouse IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR