Anti-CD38 Monoclonal Antibody [clone: AT1]

Supplier: Adipogen
ANC-187-020
102512-290EA 297.69 USD
102512-290
Anti-CD38 Monoclonal Antibody [clone: AT1]
Antibodies
Human CD38 is a type II transmembrane glycoprotein with bifunctional ectoenzyme activity catalyzing both synthesis and hydrolysis of cyclic ADP ribose. CD38 is involved in lymphocyte activation and adhesion to endothelium and has been identified as a counter-receptor of CD31.

Type: Primary
Antigen: CD38
Clonality: Monoclonal
Clone: AT1
Conjugation: Unconjugated
Epitope:
Host:
Isotype: Mouse IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR