Anti-CD37 Monoclonal Antibody [clone: IPO-24]

Supplier: Adipogen
ANC-186-020
102512-280EA 297.69 USD
102512-280
Anti-CD37 Monoclonal Antibody [clone: IPO-24]
Antibodies
Human CD37 is a member of the tetraspan family of cell surface molecules that apparantly are involved in signal transduction or transporter system complexes. CD37 is strongly expressed on B lymphocytes (sIg-positive) and weakly on T cells, neutrophils and monocytes. Expression of CD37 disappears during B cell development at the plasma cell stage.

Type: Primary
Antigen: CD37
Clonality: Monoclonal
Clone: IPO-24
Conjugation: Unconjugated
Epitope:
Host:
Isotype: Mouse IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR