Anti-CD36 Monoclonal Antibody [clone: SMO]

Supplier: Adipogen
ANC-185-820
102512-274EA 297.69 USD
102512-274
Anti-CD36 Monoclonal Antibody [clone: SMO]
Antibodies
Human CD36 is a major platelet glycoprotein also found on monocytes, macrophages and some endothelial cells. CD36 may play a role in cell adhesion as a receptor for thrombospondin. CD36 is a receptor for adhesion of malaria infected erythrocytes. CD36 is also proported to have a role in phagocytosis.

  • preservative free

Type: Primary
Antigen: CD36
Clonality: Monoclonal
Clone: SMO
Conjugation: Preservative Free
Epitope:
Host:
Isotype: Mouse IgM
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR