Anti-CD35 Monoclonal Antibody (R-PE) [clone: E11]

Supplier: Adipogen
ANC-184-050
102512-270EA 447.23 USD
102512-270
Anti-CD35 Monoclonal Antibody (R-PE) [clone: E11]
Antibodies
Human CD35 binds complement components C3b, C4b and iC3b and is expressed on erythrocytes, B cells, monocytes, granulocytes and folicular dendritic cells and on subsets of thymocytes. CD35 on erythrocytes mediates clearance of immune complexes.

Type: Primary
Antigen: CD35
Clonality: Monoclonal
Clone: E11
Conjugation: R-PE
Epitope:
Host:
Isotype: Mouse IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR