Anti-CD32 Monoclonal Antibody [clone: 7.3]

Supplier: Adipogen
ANC-181-520
102512-236EA 1903.85 USD
102512-236
Anti-CD32 Monoclonal Antibody [clone: 7.3]
Antibodies
Human CD32 exists in at least six isoforms originating from three different genes (Fc gamma RII A, B, and C). The CD32 molecule is a low affinity receptor for immune complexed IgG and has signal-transducing capabilities involved with humoral and cell-mediated immune responses.

Type: Primary
Antigen: CD32
Clonality: Monoclonal
Clone: 7.3
Conjugation:
Epitope:
Host:
Isotype: Mouse IgG1kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR