Anti-CD28 Monoclonal Antibody (Biotin) [clone: ANC28.1/5D10]

Supplier: Adipogen
ANC-177-030
102512-190EA 483.23 USD
102512-190
Anti-CD28 Monoclonal Antibody (Biotin) [clone: ANC28.1/5D10]
Antibodies
Human CD28 is an important costimulatory molecule found on all CD4+ T cells and on about half of the CD8+ T cells. T cell activities attributed to CD28 include prevention of anergy, induction of cytokine gene transcription, stabilization of cytokine mRNA and activation of CD8+ cytotoxic T lymphocytes. The ligands for CD28 identified as CD80 (B7-1) and CD86 (B7-2) are immunglobulin superfamily monomeric transmembrane glycoproteins of 60kDa and 80kDa respectively. Anti-CD28 antibodies have been shown to have a strong adjuvant effect when conjugated to an immunogen. Monovalency of the CD28 receptor for its natural ligands is essential to provide costimulation without inducing responses in the absense of TCR engagement.

Type: Primary
Antigen: CD28
Clonality: Monoclonal
Clone: ANC28.1/5D10
Conjugation: Biotin
Epitope:
Host:
Isotype: Mouse IgG1kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR