Anti-CD27 Monoclonal Antibody (FITC) [clone: M-T271]

Supplier: Adipogen
ANC-176-040
102512-182EA 350.31 USD
102512-182
Anti-CD27 Monoclonal Antibody (FITC) [clone: M-T271]
Antibodies
Human CD27 is a lymphocyte specific member of the TNF receptor family and is found primarily on peripheral blood T cells and on a subpopulation of B cells and NK cells. The ligand for CD27 is CD70, which is a member of the TNF ligand superfamily.

Type: Primary
Antigen: CD27
Clonality: Monoclonal
Clone: M-T271
Conjugation: FITC
Epitope:
Host:
Isotype: Mouse IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR