Anti-CD26 Monoclonal Antibody [clone: 202.36]

Supplier: Adipogen
ANC-175-020
102512-166EA 297.69 USD
102512-166
Anti-CD26 Monoclonal Antibody [clone: 202.36]
Antibodies
Human CD26 is a T cell activation antigen which can be associated with costimulation. The significance of the dipeptidyl peptidase IV activity remains unknown. Characterization of adenosine deaminase binding to CD26 suggests an important immunoregulatory role. Soluble CD26 serum levels are predictive for colorectal cancer.

Type: Primary
Antigen: CD26
Clonality: Monoclonal
Clone: 202.36
Conjugation: Unconjugated
Epitope:
Host:
Isotype: Mouse IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR