Anti-CD25 Monoclonal Antibody [clone: 7G7B6]

Supplier: Adipogen
ANC-174-820
102512-156EA 297.69 USD
102512-156
Anti-CD25 Monoclonal Antibody [clone: 7G7B6]
Antibodies
Expression of human CD25 (the alpha subunit of the IL-2 receptor complex) is dramatically upregulated on activated T cells, B cells, NK cells and macrophages. Antibodies to CD25 are being studied for potential use as an immunodirected therapy to selectively attack (kill) CD25 expressing cells.

  • preservative free

Type: Primary
Antigen: CD25
Clonality: Monoclonal
Clone: 7G7B6
Conjugation: Preservative Free
Epitope:
Host:
Isotype: Mouse IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR