Anti-CD22 Monoclonal Antibody (R-PE) [clone: RFB4]

Supplier: Adipogen
ANC-171-050
102512-134EA 447.23 USD
102512-134
Anti-CD22 Monoclonal Antibody (R-PE) [clone: RFB4]
Antibodies
Human CD22 expression is restricted to normal Ig+ mature B cells and neoplastic B cells. CD22 is a transmembrane glycoprotein of 140kDa with a predicted adhesion molecule function.

Type: Primary
Antigen: CD22
Clonality: Monoclonal
Clone: RFB4
Conjugation: R-PE
Epitope:
Host:
Isotype: Mouse IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR