Anti-CD21 Monoclonal Antibody [clone: BU33]

Supplier: Adipogen
ANC-170-820
102512-118EA 297.69 USD
102512-118
Anti-CD21 Monoclonal Antibody [clone: BU33]
Antibodies
Human CD21 is a receptor for complement fragments C3d, C3dg or ic3D and also EBV. CD21 is also a ligand for CD23 and plays a role in IgE synthesis.

  • preservative free

Type: Primary
Antigen: CD21
Clonality: Monoclonal
Clone: BU33
Conjugation: Preservative Free
Epitope:
Host:
Isotype: Mouse IgG1
Reactivity: Human, Primate
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR