Anti-CD18 Monoclonal Antibody (R-PE) [clone: IB4]

Supplier: Adipogen
ANC-167-050
102512-092EA 447.23 USD
102512-092
Anti-CD18 Monoclonal Antibody (R-PE) [clone: IB4]
Antibodies
Human CD18 (beta integrin) forms a heterodimer with either alphaL, alphaM, or alphaX integrins (CD11a, b or c). CD11/CD18 heterodimeric molecules are involved with cell/cell and cell/extracellular adhesion in immune and inflammatory responses.

Type: Primary
Antigen: CD18
Clonality: Monoclonal
Clone: IB4
Conjugation: R-PE
Epitope:
Host:
Isotype: Mouse IgG2a
Reactivity: Human, Primate, Rabbit
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR