Anti-CD14 Monoclonal Antibody (FITC) [clone: UCHM1]

Supplier: Adipogen
ANC-163-040
102512-050EA 350.31 USD
102512-050
Anti-CD14 Monoclonal Antibody (FITC) [clone: UCHM1]
Antibodies
Human CD14 is strongly expressed on myelomonocyte lineage cells including monocytes, macrophages and a subset of granulocytes. CD14 is a high affinity receptor for complexes of lipopolysaccharide and lipopolysaccharide binding protein. A soluble form of CD14 also binds to LPS and LBP.

Type: Primary
Antigen: CD14
Clonality: Monoclonal
Clone: UCHM1
Conjugation: FITC
Epitope:
Host:
Isotype: Mouse IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR