Anti-CD13 Monoclonal Antibody [clone: 22A5]

Supplier: Adipogen
ANC-162-820
102512-036EA 297.69 USD
102512-036
Anti-CD13 Monoclonal Antibody [clone: 22A5]
Antibodies
Human CD13 is a zinc-binding aminopeptidase-N enzyme expressed on the surface of myeloid cells. IL-4 will upregulate expression of CD13 which may play an anti-inflammatory role.

  • preservative free

Type: Primary
Antigen: CD13
Clonality: Monoclonal
Clone: 22A5
Conjugation: Preservative Free
Epitope:
Host:
Isotype: Mouse IgG2akappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR