Anti-CD11B Monoclonal Antibody (Biotin) [clone: ICRF44]

Supplier: Adipogen
ANC-159-030
102512-018EA 350.31 USD
102512-018
Anti-CD11B Monoclonal Antibody (Biotin) [clone: ICRF44]
Antibodies
Human CD11b (alpha M integrin) complexes with CD18 (beta 2 integrin) to form the complement receptor type 3 (CR3) heterodimer which binds to ICAM-1, fibrinogen, C3b and coagulation factor X. CD11b expression is restricted to leukocytes, mainly to myeloid and NK cells.

Type: Primary
Antigen: CD11B
Clonality: Monoclonal
Clone: ICRF44
Conjugation: Biotin
Epitope:
Host:
Isotype: Mouse IgG1
Reactivity: Human, Primate
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR