Anti-CD11A Monoclonal Antibody (Biotin) [clone: 38]

Supplier: Adipogen
ANC-158-030
102512-008EA 350.31 USD
102512-008
Anti-CD11A Monoclonal Antibody (Biotin) [clone: 38]
Antibodies
Human CD11a (alpha L integrin) complexes with CD18 (beta 2 integrin) to form the LFA-1 heterodimer adhesion molecule which binds to three ICAM’s 1, 2, and 3, also designated CD54, CD102 and CD50 respectively. LFA-1 is expressed on lymphocytes, monocytes and neutrophils and plays a role in cell-cell adhesion.

Type: Primary
Antigen: CD11A
Clonality: Monoclonal
Clone: 38
Conjugation: Biotin
Epitope:
Host:
Isotype: Mouse IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR