Anti-CD10 Monoclonal Antibody [clone: SN5c]

Supplier: Adipogen
ANC-157-020
102511-994EA 297.69 USD
102511-994
Anti-CD10 Monoclonal Antibody [clone: SN5c]
Antibodies
Human CD10 is a well known marker for common acute lymphoblastic leukemia antigen (CALLA). CD10 is expressed on normal early B cells in bone marrow and is also on fetal thymocytes. CD10 expression is lost as the T and B lineage cells mature and only appears again on activated germinal center B cells. CD10 is present on neutrophils. CD10 is a neutral endopeptidase and hydrolyzes peptide bonds on the amino side of hydrophobic amino acids.

Type: Primary
Antigen: CD10
Clonality: Monoclonal
Clone: SN5c
Conjugation: Unconjugated
Epitope:
Host:
Isotype: Mouse IgG1kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR