Anti-CD9 Monoclonal Antibody [clone: SN4]

Supplier: Adipogen
ANC-156-020
102511-984EA 409.85 USD
102511-984
Anti-CD9 Monoclonal Antibody [clone: SN4]
Antibodies
Human CD9 is strongly expressed on platelets, on early B cells and on activated T cells. CD9 is a member of the tetraspan family crossing the membrane four times. Both the N-terminal and C-terminal regions are on the cytoplasmic side of the plasma membrane. Studies indicate CD9 may be involved with calcium mobilization.

Type: Primary
Antigen: CD9
Clonality: Monoclonal
Clone: SN4
Conjugation: Unconjugated
Epitope:
Host:
Isotype: Mouse IgG1kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR