Anti-CD49b Monoclonal Antibody (Biotin) [clone: HAS6]

Supplier: Adipogen
ANC-155-030
102511-978EA 350.31 USD
102511-978
Anti-CD49b Monoclonal Antibody (Biotin) [clone: HAS6]
Antibodies
Human CD49b is an integrin alpha2 subunit that forms a heterodimer with beta1 integrin and functions as an adhesion molecule.

Type: Primary
Antigen: CD49b
Clonality: Monoclonal
Clone: HAS6
Conjugation: Biotin
Epitope:
Host:
Isotype: Mouse IgG2akappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR