Anti-CD8 Monoclonal Antibody [clone: 14]

Supplier: Adipogen
ANC-154-020
102511-962EA 297.69 USD
102511-962
Anti-CD8 Monoclonal Antibody [clone: 14]
Antibodies
Human CD8 is a heterodimeric protein consisting of an alpha and beta chain, each about 33kDa. CD8 is expressed on most thymocytes and on about one third of peripheral blood T cells. CD8 is a co-receptor involved in antigen recognition and binds to the alpha2 and alpha3 domain of MHC Class I molecules.

Type: Primary
Antigen: CD8
Clonality: Monoclonal
Clone: 14
Conjugation: Unconjugated
Epitope:
Host:
Isotype: Mouse IgG1kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR