Anti-CD7 Monoclonal Antibody [clone: 3A1E]

Supplier: Adipogen
ANC-152-020
102511-940EA 297.69 USD
102511-940
Anti-CD7 Monoclonal Antibody [clone: 3A1E]
Antibodies
Human CD7 is one of the earliest T lineage markers and is found on most thymocytes and peripheral blood T cells. The function of CD7 is not yet fully known, although cross-linking of CD7 with antibodies induces a T cell transmembrane calcium flux and CD7 expression is induced by ionomycin.

Type: Primary
Antigen: CD7
Clonality: Monoclonal
Clone: 3A1E
Conjugation: Unconjugated
Epitope:
Host:
Isotype: Mouse IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR